OPN on Res-Sil® C Column (packed), Restek
: Restek
This column separates the light hydrocarbons, and baseline resolves cis-2-butene from 1,3-butadiene.
- Special columns for unsaturated light hydrocarbons
- Faster separations of C1 to C4 hydrocarbons
- Res-Sil® packing replaces Porasil materials
These columns deliver faster separations of C1 to C4 hydrocarbons.
Frequently Bought Together