MXT®-1HT SimDist Column (Siltek®-treated stainless steel), Restek
Supplier: Restek
Certificates
About this item
Accurate boiling point determination for medium and heavy fractions using GC simulated distillation requires columns and phase polymers that are robust enough to withstand high temperatures without significant degradation.
- Lowest bleed for longest column lifetime
- 100% dimethyl polysiloxane phase allows easy comparisons to historical data
- Individually tested for guaranteed performance
- 7" coil Ø
- Temperature range: –60 to 450 °C
- Nonpolar phases
Maximum temperature varies depending on selected model.
Certifications: Reliably meets all ASTM D2887, D6352, D7169, D7213, and D7500 specifications.
Specifications
- Catalog No:
- 10851-186
- 10851-210
- 10851-212
- 10851-214
- 10803-430
- 10851-198
- 10850-892
- 10059-576
- 10851-200
- Description:
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- MXT-1HT SimDist
- Temperature limit:
- -60 to 380/380 °C
- -60 to 380/400 °C
- -60 to 400/430 °C
- -60 to 380/400 °C
- -60 to 430 °C
- -60 to 380/400 °C
- -60 to 430 °C
- -60 to 430/450 °C
- -60 to 400/430 °C