Siltek®-Deactivated Guard/Retention Gap Columns/Transfer Lines (fused silica), Restek
Supplier: Restek
Certificates
About this item
These guard columns and transfer lines are deactivated for superior inertness and are ideal for chlorinated pesticide analysis.
- Tested with a comprehensive test mix, to ensure high inertness
- Revolutionary deactivation process for superior inertness
- Analyze active samples accurately; ideal for chlorinated pesticide analysis (reduces endrin breakdown to less than 1%)
- Maximum temperature: 380 °C
Siltek®-deactivated gas chromatography guard columns are tested to ensure high inertness.
Frequently Bought Together