Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
 :   Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
 :  Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.
 : Triple 0.1 µm sterilized.
  • Catalog No:
  • CA45000-300
  • CA45000-302
  • CA45000-504
  • Storage temperature:
  • 2...8 °C
  • 2...8 °C
  • 2...8 °C
Product Family Options
  • Désignation
    Format du milieu
    Taille
  • Minimum Essential Medium (MEM) Alpha Medium with Earle's salts, ribonucleosides, deoxyribonucleosides, and L-glutamine
    Liquide
    500 ml
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $296.00
    $296.00
    Pack of 6 (500ml)
       $296.00
     : 10-022-CV
    Specifications
    Cat. No.
    CA45000-300
    Storage temperature
    2...8 °C
  • Minimum Essential Medium (MEM) Alpha Medium with Earle's salts without ribonucleosides, deoxyribonucleosides, and no L-glutamine
    Liquide
    500 ml
    Pack of 6 (500ml)
    Pack of 6 (500ml) - $235.00
    $235.00
    Pack of 6 (500ml)
       $235.00
     : 15-012-CV
    Specifications
    Cat. No.
    CA45000-302
    Storage temperature
    2...8 °C
  • Minimum Essential Medium (MEM) Alpha Medium, Powder with Earle's salts and L-glutamine without ribonucleosides, deoxyribonucleosides, and sodium bicarbonate
    Powder
    50 L
    Each (50l)
    Each (50l) - $121.00
    $121.00
    Each (50l)
       $121.00
     : 50-012-PC
    Specifications
    Cat. No.
    CA45000-504
    Storage temperature
    2...8 °C