Mallet with Two Replaceable Flat Plastic Ends, OR Grade, Sklar
 :   Sklar
Mallet with Two Replaceable Flat Plastic Ends, OR Grade, Sklar
 :  Sklar
Mallet with Two Replaceable Flat Plastic Ends, OR Grade, Sklar
Mallet with Two Replaceable Flat Plastic Ends, OR Grade, Sklar
Mallet with Two Replaceable Flat Plastic Ends are constructed of stainless steel and plastic
Mallet heads are 2¼" wide with a 25mm diameter. The overall weight is 0.48lbs. Industry Standard.
Product Family Options
  • Désignation
    Matière
    Longueur
  • Mallet with Two Replaceable Flat Plastic Ends
    Stainless steel
    18.4 cm (7¼")
    Each
    Each - $726.25
    $726.25
    Each
       $726.25
     : 40-5925
  • Replacement Ends For Mallet
    Plastic
    Pack of 2
    Pack of 2 - $179.56
    $179.56
    Pack of 2
       $179.56
     : 40-5926