ContactUsLinkComponent
Solid Bar Handle for Gigli Saw, OR Grade, Sklar
Catalogue # 10195-916
Fournisseur:  Sklar
undefined
Solid Bar Handle for Gigli Saw, OR Grade, Sklar
Catalogue # 10195-916
Fournisseur:  Sklar
Supplier Number:  40-5520
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Solid Bar Handle for Gigli Saw
  • Material:
    Stainless steel
  • Cat. no.:
    10195-916
  • Supplier No.:
    40-5520
Spécifications
À propos de cet article
Sklar's Gigli T Saw Handle is used in conjunction with a Gigli Wire Saw Blade
This provides an even, smooth cut. This instrument is used for sawing bones, commonly amputations, but may also be used in neurosurgery for cutting cranial flaps. The Gigli Saw is an older, manual type saw, but is still used today by surgeons.
The twisted wire is connected to two T Saw Handles and used with a pulling, back and forth motion.