Sklar's® Poole Suction Tube is an instrument used to remove large quantities of fluid from surgical sites during procedures. It is often used during abdominal procedures.
The Poole suction tube has multiple holes so as to best prevent blockages caused by fat or tissue suctioned during a procedure.
It is available in a variety of sizes, identified by the French gauge system, which measures the external diameter.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 10195-214
- 10195-506
- 10195-504
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free