PYREX® Erlenmeyer Flasks, Narrow Mouth, with Baffles, Corning
: Corning
Made of PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- For convenience, these flasks include a marking spot
- Uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance
Erlenmeyer flasks with heavy tooled rims for maximum durability. Also the baffle design promotes optimal and consistent solution agitation when used with flask shakers.
- Catalog No:
- 32645-060
- 32645-062
- 32645-056
- 32645-058
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Frequently Bought Together