Cell Detachment Solution | Avantor
Cell Detachment Solution
100 ml
Catalog # CA10120-312
Supplier:   Biolegend
Fournisseur #:  423201
Cell Detachment Solution
Catalog # CA10120-312
Supplier:  Biolegend
CAS Number:  
Cell Detachment Solution
Cell Detachment Solution
100 ml
CAS Number:  
Price
$75.30
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Accutase® is a cell detachment solution containing proteolytic and collagenolytic enzymes.
This solution is a replacement for Trypsin-EDTA solution and is useful for the detachment of cells from standard tissue culture plasticware and adhesion coated plasticware and polymers
Cell lines tested for Accutase™ application include fibroblasts, keratinocytes, vascular endothelial cells, hepatocytes, vascular smooth muscle cells, hepatocyte progenitors, primary chick embryo neuronal cells, bone marrow stem cells, adherent CHO and HNK cells, macrophages, 293 cells, L929 cells, immortalized mouse testicular germ cells, MIC5, 3T3, Vero, COS, HeLa, NT2, MG63, M24 and A375 metastatic melanoma, gliomas U251, D54, HT1080 fibrosarcoma cells, HT29 colon cancer cells, Sf9 insect cells, human embryonic stem cells, human mesenchymal stem cells and human neural stem cells.
It is a replacement for Trypsin-EDTA solution and is useful for the detachment of cells from standard tissue culture plasticware and adhesion coated plasticware and polymers.
Spécifications
Spécifications
  • Taille:
    100 ml
  • Cat. No.:
    CA10120-312
  • Enzyme name:
    Cell Detachment Solution