Model Plus: The Flower
Model Plus: The Flower, 50 cm, 50 cm, 38 cm
Catalog # 470004-728
Supplier: SI Manufacturing
Fournisseur #: 69070
Jump to:
Product Details & Documents
This colorful teaching model is designed specifically for students of all ages.
- Easy-to-Manipulate Pieces
- Guides Allow Students to Build and Learn at Own Pace
- Designed for Grades One through Eight
The model is made of resilient, non-toxic EVA foam. Removable pieces stimulate problem-solving skills and develop small motor skills.
The detailed teacher’s guide includes complete background information, reproducible worksheets, cross-curricular extension activities, assessment ideas and more.
Spécifications
- Désignation:Model Plus: The Flower
- Hauteur:38 cm
- Longueur:50 cm
- Width:50 cm