Lab Armor Beads, Chemglass
89167-124
Small Business Enterprise
- Description:Lab Armor Beads
- Size:0.75 L
- Cat. no.:89167-124
- Supplier No.:CLS-2991-075
Lab Armor™ Beads extend the life of the bath by reducing burn-out risk, eliminating corrosion and cleaning.
- Lab Armor™ bead technology saves countless hours of reworking ruined experiments
- Operating temperature of −80 to 180°C is opted for best performance
- Can be used up to 300°C without loss in thermal performance, but its fluidity may be reduced
These dry metallic beads replace water in water baths, aluminum blocks in dry baths, ice in ice buckets, and give old equipment a cool, shiny, state-of-the-art look while maintaining temperatures of -20°C to 8°C for 4 or more hours. Prevents biological or chemical contamination between samples, equipment and people that results in distorted data and non-reproducible results.