ContactUsLinkComponent
RSpec Explorer™ Digital Spectrometer
RSpec Explorer™ Digital Spectrometer
Catalog # 470220-136
undefined
RSpec Explorer™ Digital Spectrometer
Catalog # 470220-136
Supplier Number:  REX-001
Specifications
  • Description:
    RSpec Explorer™ digital spectrometer
  • Cat. no.:
    470220-136
  • Supplier No.:
    N601-RSPECEX
Specifications
About this item
The RSpec-Explorer™ digital spectrometer is simple to setup and easy to use. Position it on your lab table one or two meters from your light source. There’s no fine-tuning or adjustment necessary. The internal video camera auto-focuses and adjusts for your current light conditions.
The RSpec-Explorer™ digital classroom spectrometer is an easier and more effective way to teach visible light spectra to your physics, chemistry, and astronomy students. It is designed for demonstrating spectral sources such as gas tubes, and for measuring the spectra of a wide range of light sources, such as LEDs, street lamps, and others.
With this system, you and your students will no longer have to struggle with rainbow foil or cheap plastic spectrometers. And everyone sees the same thing at the same time. This is the only classroom spectrometer that displays live video. Set-up is simple, and no calibration is necessary. Simply position it on your lab table one or two meters from your light source. The internal video camera auto-focuses and adjusts for your current light conditions. You can be capturing your first spectrum within minutes of taking it out of the box.
RSpec Explorer™ is perfect for “sage on the stage” demonstrations, and is also an excellent lab instrument for your students to get hands-on experience.
Delivery information: The system includes a spectrometer with an internal video camera, a tabletop tripod, and one license for the award-winning RSpec Explorer™ software. This product works with all versions of Windows 10 and above, as well as with macOS.