Sklar's Pratt Uterine Dilator are a double-ended OB/GYN instrument used for progressive dilation of the uterus.
- Available in several sizes
- Dilator has two different size tips
- Straight in the middle, slightly curved at each end, with a smooth, rounded tip
These are most frequently used during intra-uterine procedures such as D&C and D&E. The size of these dilators is determined by the circumference of the tip and measured in French units, with one French unit equalling 1/3 mm.
In comparison to other intra-uterine dilators, Pratt dilators taper more gradually and are slightly lighter than Hegar dilators. They do not have a cuff to serve as a stop for depth of insertion such as featured on Hank dilators.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.