5049 Series ESD Jacket/Lab Coat, Transforming Technologies
5049 Series ESD Jacket/Lab Coat, Transforming Technologies
5049 Series ESD Jacket/Lab Coat, Transforming Technologies
5049 Series ESD Jacket/Lab Coat, Transforming Technologies
The 5049 series ESD jackets are made with more cotton than common polyester ESD jackets which makes the jackets more comfortable, breathable and softer. 5049 Fabric is made with 66% Polyester, 32% Cotton and 2% Carbon.
  • Garment maintains consistent continuity readings for up to 100 wash cycles
  • Breathable cotton poly fabric blend allows for more airflow, and is an excellent choice for very warm climates
  • Reliable panel-to-panel continuity which eliminates the danger of isolated conductors
  • Available in different lengths
  • Available with different cuff options (knitted cuff or hemmed sleeve with three snaps that adjusts for a proper fit)
Perfect for all working environments such as Electronics Manufacturing & Repair, Laboratory, Automotive, Aerospace and General Industrial Applications.
Full length option: A lab coat that goes to knee length - covers more clothing than traditional jackets.
 : Heavyweight Cotton ESD Jacket are all tested and proven to pass ANSI ESD S20.20.
  • Catalog No:
  • 10828-730
  • 10828-732
  • 10828-680
  • 10828-682
  • 10828-598
  • 10828-626
  • 10829-074
  • 10829-076
  • 10829-144
  • 10829-146
  • 10829-150
  • 10829-152
  • 10829-156
  • 10829-158
  • 10829-162
  • 10829-164
  • 10829-108
  • 10828-916
  • 10828-900
  • 10828-908
  • 10828-868
  • 10828-876
  • 10828-940
  • 10828-948
  • 10829-084
  • 10829-092
  • 10829-100
  • 10828-812
  • 10828-820
  • 10828-828
  • 10828-836
  • 10828-844
  • 10829-110
  • 10829-112
  • 10829-190
  • 10829-192
  • 10829-184
  • 10829-186
  • 10829-116
  • 10829-118
  • 10829-058
  • 10829-060
  • 10829-064
  • 10829-066
  • 10829-172
  • 10829-174
  • 10829-178
  • 10829-180
  • Model:
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • 5049
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Laboratory
  • Risk area:
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • Gender:
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • For Men
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Fabric:
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
  • 66% polyester/32% cotton, and 2% carbo
Product Family Options
  • Type
    Color
    Taille de confection
    Coupe du tenue
  • Knit, waist length
    Light blue
    S
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5402LB
    Specifications
    Cat. No.
    10828-730
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    S
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5402WH
    Specifications
    Cat. No.
    10828-732
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    M
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5403LB
    Specifications
    Cat. No.
    10828-680
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    M
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5403WH
    Specifications
    Cat. No.
    10828-682
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    L
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5404LB
    Specifications
    Cat. No.
    10828-598
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    L
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5404WH
    Specifications
    Cat. No.
    10828-626
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    XL
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5405LB
    Specifications
    Cat. No.
    10829-074
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    XL
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JWC5405WH
    Specifications
    Cat. No.
    10829-076
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    XXL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5406LB
    Specifications
    Cat. No.
    10829-144
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    XXL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5406WH
    Specifications
    Cat. No.
    10829-146
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    3XL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5407LB
    Specifications
    Cat. No.
    10829-150
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    3XL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5407WH
    Specifications
    Cat. No.
    10829-152
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    4XL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5408LB
    Specifications
    Cat. No.
    10829-156
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    4XL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5408WH
    Specifications
    Cat. No.
    10829-158
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    Light blue
    5XL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5409LB
    Specifications
    Cat. No.
    10829-162
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • Knit, waist length
    White
    5XL
    Regular
    Each
    Each - $101.00
    $101.00
    Each
       $101.00
     : JWC5409WH
    Specifications
    Cat. No.
    10829-164
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    S
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5402SPLB
    Specifications
    Cat. No.
    10829-108
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    S
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5402SPWH
    Specifications
    Cat. No.
    10828-916
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    M
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5403SPLB
    Specifications
    Cat. No.
    10828-900
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    M
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5403SPWH
    Specifications
    Cat. No.
    10828-908
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    L
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5404SPLB
    Specifications
    Cat. No.
    10828-868
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    L
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5404SPWH
    Specifications
    Cat. No.
    10828-876
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    XL
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5405SPLB
    Specifications
    Cat. No.
    10828-940
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    XL
    Regular
    Each
    Each - $84.50
    $84.50
    Each
       $84.50
     : JWC5405SPWH
    Specifications
    Cat. No.
    10828-948
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    XXL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5406SPLB
    Specifications
    Cat. No.
    10829-084
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    XXL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5406SPWH
    Specifications
    Cat. No.
    10829-092
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    3XL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5407SPLB
    Specifications
    Cat. No.
    10829-100
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    3XL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5407SPWH
    Specifications
    Cat. No.
    10828-812
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    4XL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5408SPLB
    Specifications
    Cat. No.
    10828-820
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    4XL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5408SPWH
    Specifications
    Cat. No.
    10828-828
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    Light blue
    5XL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5409SPLB
    Specifications
    Cat. No.
    10828-836
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, waist length
    White
    5XL
    Regular
    Each
    Each - $90.70
    $90.70
    Each
       $90.70
     : JWC5409SPWH
    Specifications
    Cat. No.
    10828-844
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    S
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5402SPLB
    Specifications
    Cat. No.
    10829-110
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    S
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5402SPWH
    Specifications
    Cat. No.
    10829-112
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    M
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5403SPLB
    Specifications
    Cat. No.
    10829-190
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    M
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5403SPWH
    Specifications
    Cat. No.
    10829-192
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    L
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5404SPLB
    Specifications
    Cat. No.
    10829-184
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    L
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5404SPWH
    Specifications
    Cat. No.
    10829-186
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    XL
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5405SPLB
    Specifications
    Cat. No.
    10829-116
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    XL
    Regular
    Each
    Each - $93.90
    $93.90
    Each
       $93.90
     : JLC5405SPWH
    Specifications
    Cat. No.
    10829-118
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    XXL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5406SPLB
    Specifications
    Cat. No.
    10829-058
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    XXL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5406SPWH
    Specifications
    Cat. No.
    10829-060
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    3XL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5407SPLB
    Specifications
    Cat. No.
    10829-064
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    3XL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5407SPWH
    Specifications
    Cat. No.
    10829-066
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    4XL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5408SPLB
    Specifications
    Cat. No.
    10829-172
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    4XL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5408SPWH
    Specifications
    Cat. No.
    10829-174
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    Light blue
    5XL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5409SPLB
    Specifications
    Cat. No.
    10829-178
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo
  • With snaps, knee length
    White
    5XL
    Regular
    Each
    Each - $102.00
    $102.00
    Each
       $102.00
     : JLC5409SPWH
    Specifications
    Cat. No.
    10829-180
    Model
    5049
    Garment application
    Laboratory
    Risk area
    ESD
    Gender
    For Men
    Sleeve or leg length
    Long
    Closure type
    Buttons
    Fabric
    66% polyester/32% cotton, and 2% carbo