ContactUsLinkComponent
Donor Horse Serum Gamma Irradiated
Catalogue # 10015-630
undefined
Donor Horse Serum Gamma Irradiated
Catalogue # 10015-630
Supplier Number:  DES-BTG-5XM
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    500 ml
  • Cat. no.:
    10015-630
  • Packaging:
    Nalgene bottle
  • Supplier No.:
    DES-BTG-5XM
Spécifications
À propos de cet article
Donor horse serum from RMBIO is derived from US-origin, veterinary-inspected and controlled herds
RMBIO obtains donor horse serum from closed herds that are carefully controlled and inspected by veterinarians. Horses are regularly tested for equine infectious anemia using Coggins tests. Equine serum is a cost-effective alternative to fetal bovine serum (FBS) and can be used as cell culture medium in a variety of settings. 
Equine serum is ideal for use as cell culture medium and in veterinary biologics. This product has been gamma irradiated to meet the needs of our customers.