To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Serum format:Frozen
- Description:Fetalgro Bovine Growth Serum
- Size:500 mL
- Cat. no.:10221-866
- Packaging:Nalgene bottle
- Supplier No.:FGR-BBT
Specifications
About this item
Fetalgro® is Rocky Mountain Biologicals proprietary bovine growth serum (BGS) supplemented with additives that match or outperform FBS' cell growth capabilities in many cell lines.
Each lot of Fetalgro® is tested by RMBIO's team of experienced scientists in our own state-of-the-art cell culture lab prior to releasing the product for customer use. This investment in Fetalgro's product release testing gives us the ability to find the optimal performance for specific cell lines.