Interlocking Storage Cabinets, Quantum Storage Systems
À propos de cet article
Interlocking storage cabinets have the unique ability to interlock horizontally and vertically, allowing customization based on storage needs.
- Pre-drilled holes on the backside of the cabinet allow for wall mounting
- Cabinets are made from high impact polystyrene
- Drawers are removable and will not rust or corrode
- Drawers have a recessed handle and come with a slotted label holder
Drawers have seven divider slots molded in so the drawer can be divided into multiple compartments. Clear window allows for easy identification of parts. Nine pre-configured storage system comes with drawers, clear windows and two dividers per drawer.
Drawers 11 x2 3/4 x2 1/2" Or 11 x5 5/8 x 2 1/2"
Ordering information: Additional dividers can be purchased separately.
9 Options Available Below
Product Details & Documents
Interlocking storage cabinets have the unique ability to interlock horizontally and vertically, allowing customization based on storage needs.
- Pre-drilled holes on the backside of the cabinet allow for wall mounting
- Cabinets are made from high impact polystyrene
- Drawers are removable and will not rust or corrode
- Drawers have a recessed handle and come with a slotted label holder
Drawers have seven divider slots molded in so the drawer can be divided into multiple compartments. Clear window allows for easy identification of parts. Nine pre-configured storage system comes with drawers, clear windows and two dividers per drawer.
Drawers 11 x2 3/4 x2 1/2" Or 11 x5 5/8 x 2 1/2"
Ordering information: Additional dividers can be purchased separately.
Product Family Options
Informations sur le produit
- DésignationTypeLargeurProfondeurHauteurAvailabilityPrice
- Supplier Number: QIC-64GY
- Supplier Number: QIC-83GY
- Supplier Number: QIC-4244GY
- Supplier Number: QIC-4163GY
- Supplier Number: QIC-8224GY
- Supplier Number: QIC-122GY
- Supplier Number: QIC-8143GY
- Supplier Number: QIC-12123GY
- Supplier Number: QIC-161GY
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Divider
Informations sur le produit
- Catalog #AvailabilityPrice
- Divider
Informations sur le produit
- Catalog #AvailabilityPrice
- Divider
Informations sur le produit
- Catalog #AvailabilityPrice
- Divider