Resprep® Tools for ASE® Systems, Restek
2-in-1 Filter/O-Ring Insertion Tool Kit
Catalog # 10015-286
Supplier: Restek
: 26181
Jump to:
Product Details & Documents
Resprep® tools are specialized to simplify routine chores and suitable for ASE 100/200/300.
- Use to insert filter in extraction cell or O-ring in cell cap
- Fits all extraction cells, except 1 mL size
Handle is easy to grip, and various attachments alllow for compatibility with extraction cells of all sizes.
: Includes Resprep tool handle and filter insertion attachments.
Spécifications
- Désignation:2-in-1 Filter/O-Ring Insertion Tool Kit