Florisil® PR (Bulk Adsorbent), Restek | Avantor
Florisil® PR (Bulk Adsorbent), Restek
SPE Accessory
Catalog # 10015-266
Supplier:   Restek
Fournisseur #:  26135
Florisil® PR (Bulk Adsorbent), Restek
Catalog # 10015-266
Supplier:  Restek
Florisil® PR (Bulk Adsorbent), Restek
Florisil® PR (Bulk Adsorbent), Restek
SPE Accessory
Price
$443.77
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Florisil® PR is commonly used to remove polar interferences from pesticide residues.
  • Pesticide residue grade
  • Packaged in glass containers
This bulk material is ideal for labs packing their own extraction cartridges for pesticide residue extractions.
Spécifications
  • Type:
    SPE Accessory