VWR® 60-Minute Mechanical Timer
VWR® 60-Minute Mechanical Timer
57×57 mm (2.2×2.2"), 3 minutes
Catalog # 82023-840
Supplier:   VWR International
VWR® 60-Minute Mechanical Timer
Catalog # 82023-840
Supplier:  VWR International
VWR® 60-Minute Mechanical Timer
VWR® 60-Minute Mechanical Timer
57×57 mm (2.2×2.2"), 3 minutes
Price
$73.40
New Customer Promotional Price
Log in for availability
Each Case of 20
Product Details & Documents
Dial timer has a timing capacity of 60 minutes.
  • Counts down from 60 minutes
  • Audible alarm
  • Highly visible red arrow
Specifications
  • W×H:
    57×57 mm (2.2×2.2")
  • Accuracy:
    3 minutes