McGlamry Elevator, OR Grade, Sklar
 :   Sklar
McGlamry Elevator, OR Grade, Sklar
 :  Sklar
McGlamry Elevator, OR Grade, Sklar
McGlamry Elevator, OR Grade, Sklar
McGlamry Elevators are constructed of stainless steel.
  • Industry standard
These elevators used to deglove a metatarsal head and free fused sesamoidal plates.
Product Family Options
  • Désignation
    Longueur
  • McGlamry Elevator, 11 mm
    16.5 cm (6½")
    Each
    Each - $392.91
    $392.91
    Each
       $392.91
     : 40-7480
  • McGlamry Elevator, 13 mm
    16.5 cm (6½")
    Each
    Each - $392.91
    $392.91
    Each
       $392.91
     : 40-7482
  • McGlamry Elevator, 15 mm
    16.5 cm (6½")
    Each
    Each - $392.91
    $392.91
    Each
       $392.91
     : 40-7484
  • McGlamry Elevator, 17 mm
    16.5 cm (6½")
    Each
    Each - $392.91
    $392.91
    Each
       $392.91
     : 40-7486