Mini Hohmann Elevator, OR Grade, Sklar | Avantor
Mini Hohmann Elevator, OR Grade, Sklar
Supplier:   Sklar
Mini Hohmann Elevator, OR Grade, Sklar
Supplier:  Sklar
Certificates
Mini Hohmann Elevator, OR Grade, Sklar
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mini Hohmann Elevators are constructed of stainless steel and with industry standard.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 10193-392
  • 10193-396
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Latex/Latex-free:
  • Latex-free
  • Latex-free
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Grade
    Matière
    Curvature
    Sterility
    Availability
    Price
  • Mini Hohmann Bone Elevator, 8 mm
    16.5 cm (6½")
    Premium OR
    Stainless steel
    Double ('S') Curved
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $261.93
    $261.93
    Prix promotionnel pour les nouveaux clients
    Each
       $261.93
    Supplier Number: 40-7451
    Spécifications:
    More
    Specifications
    Cat. No.
    10193-392
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
  • Mini Hohmann Elevator, 6 mm
    16.5 cm (6½")
    Premium OR-Grade
    Stainless steel
    Double ('S') Curved
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $254.09
    $254.09
    Prix promotionnel pour les nouveaux clients
    Each
       $254.09
    Supplier Number: 40-7450
    Spécifications:
    More
    Specifications
    Cat. No.
    10193-396
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free