To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Mini Tischler Universal Style Biopsy Punch
- Material:Stainless steel
- Cat. no.:10198-882
- Shape:Straight
- Supplier No.:90-7553
Spécifications
À propos de cet article
Sklar's Mini Tischler Universal Style Biopsy Punch Forceps are a non-ratcheted, double spring OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy
The Mini Tischler bite, 2.3x4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because there is typically less bleeding and pain when the smaller tissue sample is removed. It also features a single, lower tooth used to grasp difficult to reach tissue. This product is a complete instrument which consists of a handle and a detachable cannula/tip, so other universal tip styles may be used with one handle. The length of the cannula/tip is approximately 20.3cm (8").