Perforated Instrument Trays, Sklar®
Catalog # 10193-432
Supplier: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Instrument Tray, Perforated Bottom
- Dimensions:16.5×10×2.5"
- Material:Stainless steel
- Cat. no.:10193-432
- Supplier No.:10-1894
Specifications
About this item
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Perforated Trays are used for sterilizing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Perforations also prevent moisture build-up. Dome or flat lids are also available.