Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine high affinity for biotinylated target molecules with very low non specific binding. Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine fast reaction kinetics with very low non specific binding for increased throughput and precision. Applications include automated immunoassays, immunoprecipitation, and protein purification. Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation. Sera-Mag SpeedBeads and Sera-Mag Streptavidin-Coated Magnetic Particles provide a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules.
- Non-surfactant, non-protein blocking reagent provides very low non-specific binding and high binding capacity for biotinylated target molecules
- Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
- Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
- Uniform, nominal 1 μm diameter provides excellent lot-to-lot reproducibility
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA10146-022
- CA10146-018
- CA10146-020
- Binding capacity:
- 3000 - 4000 pmole/mg beads
- 3000 - 4000 pmole/mg beads
- 3000 - 4000 pmole/mg beads
- Particle size:
- nominal 1 µm Ø
- nominal 1 µm Ø
- nominal 1 µm Ø