Sera-Mag SpeedBeads® Amino-Blocked Magnetic Beads, Cytiva
Fournisseur: Cytiva
Certificats
À propos de cet article
Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.
- Non-surfactant, non-protein blocking reagent provides very low non-specific binding of proteins from the sample matrix
- Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
- Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
- Uniform, nominal 1 μm diameter provides excellent lot-to-lot reproducibility
Sera-Mag SpeedBeads Amine-Blocked Particles combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics. Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays.
Spécifications
- Catalog No:
- CA10146-014
- CA10146-016
- CA10146-012
- Particle size:
- nominal 1 µm diameter
- nominal 1 µm diameter
- nominal 1 µm diameter