Sera-Mag SpeedBeads® Protein A/G Magnetic Beads, Cytiva
Supplier: Cytiva
Certificates
About this item
Sera-Mag SpeedBeads Protein A/G Magnetic Particles enable magnetic isolation of IgA and IgG proteins in a single isolation. Sera-Mag SpeedBeads Protein A/G Magnetic Particles provide a fast and convenient method for manual and automated magnetic isolation of affinity bound antibodies or antigens to Protein A/G. These particles can be used for isolating antibodies from serum, cell culture supernatants, or ascites, and for immunoprecipitation and co-immunoprecipitation of antigens from cell or tissue extracts.
- Manual and automated magnetic isolation of IgA and IgG proteins is accomplished in a single isolation instead of running two separate isolations
- Protein A/G binding sites allows use of a broad range of antibody species and sub-classes
- Low non-specific binding for fast and clean purification
- Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
- Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
- Uniform, nominal 1 μm diameter provides excellent lot-to-lot reproducibility
Specifications
- Catalog No:
- CA10146-046
- CA10146-044
- CA10146-042
- Particle size:
- nominal 1 µm diameter
- nominal 1 µm diameter
- nominal 1 µm diameter