Krey Hook, Sklar
Krey Hook, Sklar
Krey Hook, Double Action, 20.3 cm (8"), Stainless steel
Catalog # 10185-492
Supplier:   Sklar
Supplier #:  93-8007
Krey Hook, Sklar
Catalog # 10185-492
Supplier:  Sklar
Krey Hook, Sklar
Krey Hook, Sklar
Krey Hook, Double Action, 20.3 cm (8"), Stainless steel
Price
$209.39
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Krey Hooks are construced of stainless steel.
Industry standard.
Specifications
  • Désignation:
    Krey Hook, Double Action
  • Longueur:
    20.3 cm (8")
  • Matière:
    Stainless steel