Sklar Kelly Retractors are available in two distinct styles
It is available with various blade lengths and can therefore retract several layers of tissue if necessary. This retractor is often confused with the Richardson retractor, however, the Kelly retractor features longer blades. Sklar offers this Kelly retractor with a grip handle that is hollow and features finger ridges and a slight hook at the end for a secure grip and improved comfort.
This Kelly Retractor is a manual, right angle, general retractor used for wound exposure.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 10197-714
- 10197-716
- 10197-710
- 10197-712
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free