ContactUsLinkComponent
Merit™ Sims Uterine Sound, Physician Grade, Sklar
Catalog # 10185-324
Supplier:  Sklar
undefined
Merit™ Sims Uterine Sound, Physician Grade, Sklar
Catalog # 10185-324
Supplier:  Sklar
Supplier Number:  98-384
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Merit™ Sims Uterine Sound, Plain
  • Length:
    31.7 cm (12½")
  • Material:
    Stainless steel, silver plated
  • Cat. no.:
    10185-324
  • Supplier No.:
    98-384
Specifications
About this item
The Merit™ Sims Uterine Sound is a malleable, silver-plated OB/GYN instrument used for probing and dilating the uterus as well as measuring the length of the cervical canal.
It is very similar to the Simpson uterine sound. The instrument is available with or without graduated centimeter markings. This product is straight, but malleable and can be curved as necessary. The handle is a short, smooth thumb grip that features three vertical grooves for improved grip.