Snook Hook, Sklar
Product Details & Documents
These are made of stainless steel.
- Reusable
- Non-sterile
- Latex-Free
Premium OR-grade.
Spécifications
- Désignation:Snook Hook
- Tip Configuration:Blunt
- Working Surface Style:Rounded
- Tip Dimensions:1/4"
- Handle:Solid Flat