ContactUsLinkComponent
Merit™ Buck Neuro Hammer, Physician Grade, Sklar
Catalogue # 10184-686
Fournisseur:  Sklar
undefined
Merit™ Buck Neuro Hammer, Physician Grade, Sklar
Catalogue # 10184-686
Fournisseur:  Sklar
Supplier Number:  97-021
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Merit™ Buck Neuro Hammer
  • Length:
    17.8 cm (7")
  • Material:
    Stainless steel
  • Cat. no.:
    10184-686
  • Supplier No.:
    97-021
Spécifications
À propos de cet article
The Merit™ Buck Neuro Hammer is a diagnostic instrument to evaluate myotatic and cutaneous reflexes.
This can often lead to detection of other neurological abnormalities in a patient. The double head consists of two rubber tips of different sizes. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This hammer features a brush and needle, which are stored in the handle of the instrument.