Ready Engineered Complete Pallet Shipping Solutions, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
À propos de cet article
Complete Pallet Shipping Solutions are designed to meet the unique transport requirements of pharmaceuticals, biologics, vaccines, and other high value life science products.
- Pre-qualified for refrigerated, frozen, and CRT temperatures
- Optimized size
Featuring more temperature options, fewer packing components, and easy assembly, these pallet solutions are designed to meet the unique transport requirements of pharmaceuticals, biologics, vaccines, and other high value life science products. Dimensions have been optimized for narrow-body aircraft, wide-body aircraft, and North American trucks.
Ordering information: For Tall US Pallet as a Payload-Seasonal, order (1) Pallet Shell, and the following Refrigerant Sleeve(s) depending on desired temperature coverage/duration. For <-20°C, 48 hrs duration or <-15°C, 120 hrs duration, order (1) SPS-FS. For 2-8°C, 96 hrs duration or 1-10°C, 144 hrs duration, order (1) SPS-RSR and (1) SPS-RSF. For 15-25°C, 72 hrs duration or 10-30°C, 120 hrs duration, order (1) SPS-CRT.
For Universal 7-day protection for US 48” x 40” Pallet as a Payload, order (1) pallet shell GPS-RP-PL, (16) 6040ASLEVR sleeves and (12) 6040WSLEVF sleeves.
For 5 and 7-day seasonal protection, the GPS-RPS (summer) and GPS-RPW (winter) models include all necessary components for the stated performance and duration.
For Universal 7-day protection for US 48” x 40” Pallet as a Payload, order (1) pallet shell GPS-RP-PL, (16) 6040ASLEVR sleeves and (12) 6040WSLEVF sleeves.
For 5 and 7-day seasonal protection, the GPS-RPS (summer) and GPS-RPW (winter) models include all necessary components for the stated performance and duration.
Frequently Bought Together