BathBrite™ Cleaner
BathBrite™
Supplier: Decon Labs
Supplier #: 3408
Jump to:
Product Details & Documents
BathBrite™ is designed for use in water baths, incubators, circulators, and other similar temperature controlled lab equipment.
Supplied in a 237 ml (8 oz.) bottle
- Bottle features a flip-top cap for easy dispensing
Adding 10 to 15 drops of BathBrite™ per gallon of water will keep water clear and fresh, and extend the time between water replacement.
Documents
Certificates
Search by Lot NumberSpecifications
- Désignation:BathBrite™