ContactUsLinkComponent
Optical Lens Cleaner, FoxWest Sales
Optical Lens Cleaner, FoxWest Sales
Catalogue # 10611-616
Fournisseur:  Decon Labs
undefined
Optical Lens Cleaner, FoxWest Sales
Catalogue # 10611-616
Fournisseur:  Decon Labs
Supplier Number:  1100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Optical lens cleaner, 60 ml (2 oz.)
  • Size:
    6 lenses/pack, 8 packs/case
  • Cat. no.:
    10611-616
  • Supplier No.:
    1100
Spécifications
À propos de cet article
Optical lens cleaner is a gentle cleaning solution that can be used to clean all optical glass surfaces including microscope optics, absorption cells, and lenses.
  • Optical lens cleaner can be used to clean all optical glass surfaces
  • Contains no abrasives or silicones
  • Safe to use with cemented lenses
This cleaner contains no abrasives or silicones so it will not etch, scratch, or streak.