Army Pattern Chisel, OR Grade, Sklar
 :   Sklar
Army Pattern Chisel, OR Grade, Sklar
 :  Sklar
Army Pattern Chisel, OR Grade, Sklar
Army Pattern Chisel, OR Grade, Sklar
Army Pattern Chisels are Stainless Steel and Industry Standard.
Product Family Options
  • Désignation
    Longueur
    Matière
    Tip Dimensions
    Curvature
  • Army Pattern Chisel
    6½"
    Stainless steel
    6 mm
    Straight
    Each
    Each - $315.16
    $315.16
    Each
       $315.16
     : 40-6106
  • Army Pattern Chisel
    6½"
    Stainless steel
    12 mm
    Straight
    Each
    Each - $363.77
    $363.77
    Each
       $363.77
     : 40-6112
  • Army Pattern Chisel
    6½"
    Stainless steel
    18 mm
    Straight
    Each
    Each - $329.69
    $329.69
    Each
       $329.69
     : 40-6118