Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Sterility:Non sterile
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Description:US Army Retractor
- Length:20.9 cm (8¼")
- Material:Stainless steel
- Tip Configuration:Blade
- Curvature:Straight
- Cat. no.:10192-346
- Grade:Premium OR-Grade
- Supplier No.:60-1085
The US Army Retractor is used to expose surface layers of skin, commonly in plastic surgery procedures.
It is a double-ended retractor with a fenestrated handle
Censitrac Ready products are industry standard and constructed of stainless steel.
The blades at each end are angled 90° and also have a slightly curved, rounded lip. The blades also face in the same direction. US Army Retractors come in sets of 2.