Cables, Huber
Cables, Huber
Supplier:   Huber
Cables, Huber
Supplier:  Huber
Certificates
Cables, Huber
1 Options Available Below
Product Details & Documents
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Cable CC/RS232 (PC)
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Cable compatible control, CC AIF / POKO
    3 m (10 ft.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $401.16
    $401.16
    Each
       $401.16
    Minimum order quantity is 1
    Supplier Number: 6218
  • Cable compatible control, CC RS232 / PC
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Connecting cable, Pilot ONE, Mini–USB / PC, USB
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable RS232 (15 <> 9 pin)
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable Unipump / Unistat
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, AIF / open end
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, CP AIF / open end
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, CP RS485 / open end
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, ECS / open end
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, LEVEL / open end
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, POKO / alarm Unistat
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control cable, RS232 (9 <> 9 pin)
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Control panel extension cable for Pilot ONE
    3 m (10 ft.)
    Discontinued
    Supplier Number: -
  • Extension cable with plug and connection
    3 m (10 ft.)
    Discontinued
    Supplier Number: -