Starfish Slides | Avantor
Starfish Slides
Supplier:   Avantor
Starfish Slides
Supplier:  Avantor
Certificates
Starfish Slides
5 Options Available Below
Product Details & Documents
Explore Starfish Characteristics Microscopically
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Starfish are interesting animals and are members of the phylum Echinodermata
We offer individual starfish slides of:
▪ Starfish Arm - Cross sections through two regions of the arm.
▪ Starfish, Pedicellariae - Stalked type. Isolated organs.
▪ Starfish, Sperm - General structure of starfish sperm.
▪ Starfish, Testis - Displays structure of starfish testis.
▪ Starfish, Ovary - General structure of starfish ovary.
This phylum also includes sea urchins, sea cucumbers, and sand dollars, among others. Most echinoderms are able to grow back limbs and organs if they’ve been damaged or destroyed. Echinoderms have bilaterial symmetry (like humans) during early development but then shift to radial symmetry as they grow to maturity.
Product Family Options
Informations sur le produit
  • Désignation
    Orientation
    Stain
    Availability
    Price
  • Sperm - General structure
    Smear
    Hematoxylin
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.35
    $10.35
    Each
       $10.35
  • Arm - Cross sections through two regions
    Section
    Mallory
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13.29
    $13.29
    Each
       $13.29
  • Ovary - General structure
    Section
    Hematoxylin and Eosin
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17.77
    $17.77
    Each
       $17.77
  • Stalked Pedicellariae - Isolated organs
    Whole mount
    Fast Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18.70
    $18.70
    Each
       $18.70
  • Testis – General Structure
    Section
    Hematoxylin and Eosin
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $16.93
    $16.93
    Each
       $16.93