Scypha Slides
Scypha Slides
Supplier:   Avantor
Scypha Slides
Supplier:  Avantor
Certificates
Scypha Slides
6 Options Available Below
Product Details & Documents
Use Scypha slides to teach about marine life or compare and contrast animal body types
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Scypha are small, tube shaped marine calcarious sponges. They are sedentary and attach in colonies to rocks and other non-moving material in shallow seawater. Scypha typically have a diploblastic body and show a syconoid type of canal system.
We offer individual Scypha slides of:
Scypha grantia (cross section) hematoxylin & eosin- Ascon type sponge. Decalcified and stained to show the general structure of the canals and body wall.
Scypha (median- longitudinal section) h&e- Section through body and osculum. It highlights the relationship between incurrent and radial canals.
Scypha (longitudinal section) hematoxylin & eosin- Near median section shows relationship of incurrent and radial canals.
Scypha, (cross section & longitudinal section) hematoxylin & eosin- Demonstrates general structure of canals and body wall.
Scypha, thick section (cross section) hematoxylin & eosin- Thick section. Shows spicules and their relationship to canal systems.
Scypha, Spicule Strew (whole mount)- Typical forms of calcarious spicules. Unstained.
Product Family Options
Product Information
  • Désignation
    Orientation
    Stain
    Availability
    Price
  • Scypha grantia, (cs)
    Section
    Hematoxylin and Eosin
    Log in for availability
    Log in for availability
    Each
    Each - $14.40
    $14.40
    Each
       $14.40
  • Scypha, (cs & ls)
    Cross section & longitudinal section
    Hematoxylin and Eosin
    Log in for availability
    Log in for availability
    Each
    Each - $27.57
    $27.57
    Each
       $27.57
  • Scypha, (cs)
    Cross section
    Hematoxylin and Eosin
    Log in for availability
    Log in for availability
    Each
    Each - $24.07
    $24.07
    Each
       $24.07
  • Scypha, (ls)
    Longitudinal section
    Hematoxylin and Eosin
    Log in for availability
    Log in for availability
    Each
    Each - $15.40
    $15.40
    Each
       $15.40
  • Scypha, (mls)
    Medial longitudinal section
    Hematoxylin and Eosin
    Log in for availability
    Log in for availability
    Each
    Each - $18.90
    $18.90
    Each
       $18.90
  • Scypha, Spicule Strew (wm)
    Whole mount
    Unstained
    Log in for availability
    Log in for availability
    Each
    Each - $8.70
    $8.70
    Each
       $8.70