Simpson Uterine Sound, OR Grade, Sklar
Fournisseur: Sklar
Certificats
À propos de cet article
Thiese are malleable, OB/GYN instruments used for probing and dilating the uterus, as well as measuring the length of the cervical canal.
- Made of silver plated stainless steel
- Round and thin with a small nodule at the tip and a thumb grip handle
Very similar to the Sims uterine sound.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together