ESD Modular Trays, Quantum Storage Systems | Avantor
ESD Modular Trays, Quantum Storage Systems
ESD Modular Trays, Quantum Storage Systems
Certificates
ESD Modular Trays, Quantum Storage Systems
ESD Modular Trays, Quantum Storage Systems
2 Options Available Below
Product Details & Documents
These trays are ESD and comply with ANSI/ESD standards for the use in PC board assembly and handling.
  • Withstand temperatures of −40 °F to +350 °F
  • Trays are resistant to abrasion and chemicals
  • Trays are permanently conductive, properties are unaffected by washing in steam or water
All trays are manufactured of fiberglass reinforced thermoset material, allowing up to 200 lb. weight capacity when evenly distributed.
Product Family Options
Informations sur le produit
  • Désignation
    L×W×H
    Availability
    Price
  • ESD Trays; Conductive
    18 × 26 × 1 1/8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $192.77
    $192.77
    Each
       $192.77
    La quantité minimale de commande est de 1
    Supplier Number: 1826ESD
  • ESD Trays; Conductive
    20 × 26 × 1 1/2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $357.06
    $357.06
    Each
       $357.06
    La quantité minimale de commande est de 1
    Supplier Number: 2026ESD