Micro-Cut Wittner Uterine Biopsy Punch, OR Grade, Sklar
10199-186
: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Micro-Cut Wittner Uterine Biopsy Punch Straight 4mm x 8mm Bite
- Material:Stainless steel
- Cat. no.:10199-186
- Supplier No.:90-8900
Sklar® Micro-Cut Wittner Uterine Biopsy Punch is a reusable, stainless steel, OR grade punch.
- Used in general surgery
- Obstetrics and gynecology Physician