Micro-Cut Eppendorfer Biopsy Punch, OR Grade, Sklar
10196-414
: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Stainless Steel
- Length:12"
- Material:Stainless steel
- Tip Configuration:Semi Elliptical/Semi Oval Bite
- Curvature:Straight
- Cat. no.:10196-414
- Supplier No.:90-8010
Sklar® Micro-Cut Eppendorfer Biopsy Punch is a reusable, stainless steel, OR grade punch.
- Used in general surgery
- Obstetrics and gynecology Physician
This product is straight, semi elliptica/semi oval bite tip, with finger ring handle style. Instrument measures 12” in length with a 8” shaft length.