Wire Shelving Systems with Conductive Bins, Quantum Storage Systems
These shelve units come standard with conductive bins, conductive split sleeves and a drag chain to be attached to the bottom shelf helping to ground the shelving unit.
- Eliminate the build up of a conductive charge
- Protect the electronic components being stored
ESD-safe.
: Supplied with black bins.
Frequently Bought Together