VWR® Built In Undercounter Refrigerators with Natural Refrigerants
Supplier: VWR International
Certificates
About this item
These undercounter laboratory refrigerators provide lab quality cold storage in a space-saving design.
- Integrated temperature alarms
- Keyed door locks
- Forced draft circulation
- Precise microprocessor temperature control
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 10819-880, 10819-884, 76210-110, 10819-882, 76210-108, 76210-106, 76210-112, 10819-886
Ordering information: All units have a two-year parts and labor warranty. Select models come with an additional three-year compressor parts warranty. (Please refer to unit datasheets to confirm).
Specifications
- Catalog No:
- 76210-112
- 76210-110
- 10819-882
- 76210-108
- 76210-106
- 10819-880
- 10819-886
- 10819-884
- Model:
- HCUCBI0404SSLH
- HCUCBI0404ADLH
- HCUCBI0404G
- HCUCBI0404GLH
- HCUCBI0404LH
- HCUCBI0404
- HCUCBI0404SS
- HCUCBI0404AD
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76210-112
- 76210-110
- 10819-882
- 76210-108
- 76210-106
- 10819-880
- 10819-886
- 10819-884
- Shelves/drawers:
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- 2 adjustable shelves, 1 fixed shelf
- Door type:
- 1 stainless steel swing door, hinge left
- 1 glass swing door, hinge left
- 1 glass swing door, hinge right
- 1 glass swing door, hinge left
- 1 solid swing door, hinge left
- 1 solid swing door, hinge right
- 1 stainless steel swing door, hinge right
- 1 glass swing door, hinge right
- Electrical:
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- 115 V, 60 Hz, 1.74 A
- Shipping weight:
- 61 kg (135 lbs.)
- 68 kg (150 lbs.)
- 58 kg (127 lbs.)
- 58 kg (127 lbs.)
- 62 kg (137 lbs.)
- 62 kg (137 lbs.)
- 61 kg (135 lbs.)
- 68 kg (150 lbs.)
- Environmental certification:
- CFC Free
- CFC Free
- CFC Free
- CFC Free
Specifications
Related Information
Product Information
Frequently Bought Together