Vernier® Digital Sensor Cable
Vernier digital sensor cable
Catalog # CA470219-300
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Supplier #: MDC-BTD
Vernier® Digital Sensor Cable
Catalog # CA470219-300
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
This cable connects Vernier digital sensors to the digital (DIG) port on a Vernier interface.
Caution: Not compatible with the Photogate.
Specifications
- Désignation:Vernier digital sensor cable