Vernier® Digital Sensor Extension Cable
Vernier digital sensor extension cable, 2m
Catalog # CA470219-278
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: EXT-BTD
Vernier® Digital Sensor Extension Cable
Catalog # CA470219-278
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
The Digital Sensor Extension Cable can be used with any Vernier sensor that connects to a digital channel of one of our interfaces. The port will be labeled DIG1 or DIG2.
Spécifications
- Désignation:Vernier digital sensor extension cable, 2m