Vernier® Analog Sensor Extension Cable
Vernier analog sensor extension cable, 2 m
Catalog # CA470219-276
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: EXT-BTA
Vernier® Analog Sensor Extension Cable
Catalog # CA470219-276
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
Most of the Vernier sensors have cables that are 1.5 m long.
There are times, however, that users may wish to have a longer cable. The Analog Sensor Extension Cable can be used with any Vernier sensor that connects to an analog channel labeled CH1, CH2, CH3, or CH4. Each cable is 2 m in length.
Spécifications
- Désignation:Vernier analog sensor extension cable, 2 m