To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Sklar Black™ Babcock Forceps
- Length:9½"
- Material:Stainless Steel with Reduced Reflectivity Polymer Coating
- Curvature:Straight
- Cat. no.:10199-106
- Supplier No.:90-2295
Sklar Black™ Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue in laser procedures
They are frequently used with intestinal and laparotomy procedures. Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.